Anti-IAA Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Lieferant: Bioss
BS-0902R-A680
BOSSBS-0902R-A680EA 890 EUR
BOSSBS-0902R-A680
Anti-IAA Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
Indole-3-acetic acid, also known as IAA, is a heterocyclic compound that is an phytohormones called auxins. This colourless solid is probably the most important plant auxin. The molecule is derived from indole, containing a carboxymethyl group (acetic acid). IAA has many different effects, as all auxins do, such as inducing cell elongation and cell division with all subsequent results for plant growth and development. There are less expensive and metabolically stable synthetic auxin analogs on the market for use in horticulture, such as indole-3-butyric acid (IBA) and 1-naphthaleneacetic acid (NAA).

Type: Primary
Antigen: IAA
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range:
Host: Rabbit
Isotype: IgG
Reactivity: Others
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR