Anti-BrdU Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Lieferant: Bioss
BS-0917R-A488
BOSSBS-0917R-A488EA 893 EUR
BOSSBS-0917R-A488
Anti-BrdU Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antikörper
Bromodeoxyuridine is a synthetic nucleoside that is an analogue of thymidine. BrdU is incorporated into DNA during S-phase of a cell cycle. It is commonly used in the detection of proliferating cells in living tissues
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: BrdU
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Others