Anti-BrdU Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Lieferant: Bioss
BS-0917R-A555
BOSSBS-0917R-A555EA 893 EUR
BOSSBS-0917R-A555
Anti-BrdU Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antikörper
Bromodeoxyuridine is a synthetic nucleoside that is an analogue of thymidine. BrdU is incorporated into DNA during S-phase of a cell cycle. It is commonly used in the detection of proliferating cells in living tissues

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: BrdU
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Others
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR