Anti-MTNR1B Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Lieferant: Bioss
BS-0963R-A488
BOSSBS-0963R-A488EA 901 EUR
BOSSBS-0963R-A488
Anti-MTNR1B Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antikörper
High affinity receptor for melatonin. Likely to mediates the reproductive and circadian actions of melatonin. The activity of this receptor is mediated by pertussis toxin sensitive G proteins that inhibit adenylate cyclase activity.

Recommended Dilutions: IF(IHC-P): 1:50-200; IF(ICC)(1:50-200

Type: Primary
Antigen: MTNR1B
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Bovine
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR