Anti-CD38 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Lieferant: Bioss
BS-0979R-A680
BOSSBS-0979R-A680EA 890 EUR
BOSSBS-0979R-A680
Anti-CD38 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
synthesises the second messagers cyclic ADP-ribose and nicotinate-adenine dinucleotide phosphate, the former a second messenger for glucose-induced insulin secretion. Also has cADPr hydrolase activity (By similarity).
Type: Primary
Antigen: CD38
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 101-200/304
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat