Anti-CD38 Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Lieferant: Bioss
BS-0979R-A750
BOSSBS-0979R-A750EA 890 EUR
BOSSBS-0979R-A750
Anti-CD38 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
synthesises the second messagers cyclic ADP-ribose and nicotinate-adenine dinucleotide phosphate, the former a second messenger for glucose-induced insulin secretion. Also has cADPr hydrolase activity (By similarity).

Type: Primary
Antigen: CD38
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 101-200/304
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR