Anti-CD38 Rabbit Polyclonal Antibody (Cy3®)

Lieferant: Bioss
BS-0979R-CY3
BOSSBS-0979R-CY3EA 900 EUR
BOSSBS-0979R-CY3
Anti-CD38 Rabbit Polyclonal Antibody (Cy3®)
Antikörper
Synthesizes the second messagers cyclic ADP-ribose and nicotinate-adenine dinucleotide phosphate, the former a second messenger for glucose-induced insulin secretion. Also has cADPr hydrolase activity (By similarity).

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CD38
Clonality: Polyclonal
Clone: 0
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR