Anti-CD38 Rabbit Polyclonal Antibody (Cy5.5®)
Lieferant: Bioss
BS-0979R-CY5.5
BOSSBS-0979R-CY5.5EA 900 EUR
BOSSBS-0979R-CY5.5
Anti-CD38 Rabbit Polyclonal Antibody (Cy5.5®)
Antikörper
Synthesizes the second messagers cyclic ADP-ribose and nicotinate-adenine dinucleotide phosphate, the former a second messenger for glucose-induced insulin secretion. Also has cADPr hydrolase activity (By similarity).
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: CD38
Clonality: Polyclonal
Clone: 0
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen