Anti-TFRC Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Lieferant: Bioss
BS-0988R-A350
BOSSBS-0988R-A350EA 893 EUR
BOSSBS-0988R-A350
Anti-TFRC Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antikörper
Cellular uptake of iron occurs via receptor-mediated endocytosis of ligand-occupied transferrin receptor into specialized endosomes. Endosomal acidification leads to iron release. The apotransferrin-receptor complex is then recycled to the cell surface with a return to neutral pH and the concomitant loss of affinity of apotransferrin for its receptor. Transferrin receptor is necessary for development of erythrocytes and the nervous system (By similarity). A second ligand, the heditary hemochromatosis protein HFE, competes for binding with transferrin for an overlapping C-terminal binding site.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: TFRC
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat