Anti-CTLA4 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Lieferant: Bioss
BS-10006R-HRP
BOSSBS-10006R-HRPEA 901 EUR
BOSSBS-10006R-HRP
Anti-CTLA4 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
Inhibitory receptor acting as a major negative regulator of T-cell responses. The affinity of CTLA4 for its natural B7 family ligands, CD80 and CD86, is considerably stronger than the affinity of their cognate stimulatory coreceptor CD28.
Recommended Dilutions: Western Blot: 1:100-1000
Type: Primary
Antigen: CTLA4
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Bestellen