Anti-DIM-7 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Lieferant: Bioss
BS-10015R-A488
BOSSBS-10015R-A488EA 893 EUR
BOSSBS-10015R-A488
Anti-DIM-7 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antikörper
DIM-7
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: DIM-7
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Other Species