Anti-DIM-7 Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Lieferant: Bioss
BS-10015R-A647
BOSSBS-10015R-A647EA 893 EUR
BOSSBS-10015R-A647
Anti-DIM-7 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antikörper
DIM-7

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: DIM-7
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Other Species
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR