Anti-DIM-7 Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Lieferant: Bioss
BS-10015R-A750
BOSSBS-10015R-A750EA 890 EUR
BOSSBS-10015R-A750
Anti-DIM-7 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
DIM-7.

Type: Primary
Antigen: DIM-7
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 401-500/1049
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR