Anti-DIM-7 Rabbit Polyclonal Antibody (Cy3®)

Lieferant: Bioss
BS-10015R-CY3
BOSSBS-10015R-CY3EA 900 EUR
BOSSBS-10015R-CY3
Anti-DIM-7 Rabbit Polyclonal Antibody (Cy3®)
Antikörper
DIM-7

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: DIM-7
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Other Species
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR