Anti-DIM-7 Rabbit Polyclonal Antibody (Cy5®)
Lieferant: Bioss
BS-10015R-CY5
BOSSBS-10015R-CY5EA 900 EUR
BOSSBS-10015R-CY5
Anti-DIM-7 Rabbit Polyclonal Antibody (Cy5®)
Antikörper
DIM-7
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: DIM-7
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Other Species