Anti-alpha s1 Casein Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Lieferant: Bioss
BS-10033R-HRP
BOSSBS-10033R-HRPEA 900 EUR
BOSSBS-10033R-HRP
Anti-alpha s1 Casein Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
Alpha s1 Casein

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: [delete]
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bovine
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR