Anti-EIF5 Rabbit Polyclonal Antibody (Cy3®)

Lieferant: Bioss
BS-10035R-CY3
BOSSBS-10035R-CY3EA 893 EUR
BOSSBS-10035R-CY3
Anti-EIF5 Rabbit Polyclonal Antibody (Cy3®)
Antikörper
Catalyzes the hydrolysis of GTP bound to the 40S ribosomal initiation complex (40S.mRNA.Met-tRNA[F].eIF-2.GTP) with the subsequent joining of a 60S ribosomal subunit resulting in the release of eIF-2 and the guanine nucleotide. The subsequent joining of a 60S ribosomal subunit results in the formation of a functional 80S initiation complex (80S.mRNA.Met-tRNA[F]).

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: EIF5
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR