Anti-SP-C Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Lieferant: Bioss
BS-10067R-A680
BOSSBS-10067R-A680EA 899 EUR
BOSSBS-10067R-A680
Anti-SP-C Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
Pulmonary surfactant associated proteins promote alveolar stability by lowering the surface tension at the air-liquid interface in the peripheral air spaces.

Type: Primary
Antigen: SP-C
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 24-58/197
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR