Anti-SLC7A9 Rabbit Polyclonal Antibody (Cy5®)

Lieferant: Bioss
BS-10085R-CY5
BOSSBS-10085R-CY5EA 1350 EUR
BOSSBS-10085R-CY5
Anti-SLC7A9 Rabbit Polyclonal Antibody (Cy5®)
Antikörper
Involved in the high-affinity, sodium-independent transport of cystine and neutral and dibasic amino acids (system b(0,+)-like activity). Thought to be responsible for the high-affinity reabsorption of cystine in the kidney tubule.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: SLC7A9
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR