Anti-CXCR4 Rabbit Polyclonal Antibody
Lieferant: Bioss
BS-1011R
BOSSBS-1011REA 752 EUR
BOSSBS-1011R
Anti-CXCR4 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: CXCR4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Bestellen