Anti-IKK Alpha + IKK beta Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Lieferant: Bioss
BS-10123R-A555
BOSSBS-10123R-A555EA 893 EUR
BOSSBS-10123R-A555
Anti-IKK Alpha + IKK beta Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antikörper
IKK Alpha/IKK beta is a member of the IKK complex which is composed of IKK alpha, IKK beta, IKK gamma and IKAP. Phosphorylation of I-Kappa-B on a serine residue by the IKK complex frees NF-kB from I-Kappa-B and marks it for degradation via ubiquination. IKK beta has been shown to activate NF-kB and phosphorylate IKB alpha and beta. Phosphorylation of 2 sites at the activation loop of IKK beta is essential for activation of IKK by TNF and IL1. Once activated, IKK beta autophosphorylates which in turn decreases IKK activity and prevents prolonged activation of the inflammatory response. Additionally, IKK beta activity can also be regulated by MEKK1.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: IKK Alpha + IKK beta
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat