Anti-LAT Tyr161 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Lieferant: Bioss
BS-10127R-A680
BOSSBS-10127R-A680EA 917 EUR
BOSSBS-10127R-A680
Anti-LAT Tyr161 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
Required for TCR (T-cell antigen receptor)- and pre-TCR-mediated Signalling, both in mature T-cells and during their development. Involved in FCGR3 (low affinity immunoglobulin gamma Fc region receptor III)-mediated Signalling in natural killer cells and FCER1 (high affinity immunoglobulin epsilon receptor)-mediated Signalling in mast cells. Couples activation of these receptors and their associated kinases with distal intracellular events such as mobilization of intracellular calcium stores, PKC activation, MAPK activation or cytoskeletal reorganization through the recruitment of PLCG1, GRB2, GRAP2, and other Signalling molecules.
Type: Primary
Antigen: LAT Tyr161
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen