Anti-LAT Rabbit Polyclonal Antibody

Lieferant: Bioss
BS-10128R
BOSSBS-10128REA 743 EUR
BOSSBS-10128R
Anti-LAT Rabbit Polyclonal Antibody
Antikörper
Linker for activation of T cells (LAT) is an integral membrane protein (36 to 38 kD), which plays an important role in linking engagement of the TCR to the biochemical events of T cell activation. LAT antibody stains thymus and peripheral lymphoid tissues such as T cell areas in lymph nodes and spleen. LAT is expressed in T lymphocytes in interstitial spaces, platelets and megakaryocytes. LAT is not expressed in B cells, macrophages, plasma cells, monocytes, epithelial histiocytes and dendritic cells.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: LAT
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR