Anti-APOD Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Lieferant: Bioss
BS-10130R-A555
BOSSBS-10130R-A555EA 893 EUR
BOSSBS-10130R-A555
Anti-APOD Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antikörper
APOD occurs in the macromolecular complex with lecithin-cholesterol acyltransferase. It is probably involved in the transport and binding of bilin. Appears to be able to transport a variety of ligands in a number of different contexts.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: APOD
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR