Anti-SCNN1D Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Lieferant: Bioss
BS-10141R-A488
BOSSBS-10141R-A488EA 893 EUR
BOSSBS-10141R-A488
Anti-SCNN1D Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antikörper
Sodium permeable non-voltage-sensitive ion channel inhibited by the diuretic amiloride. Mediates the electrodiffusion of the luminal sodium (and water, which follows osmotically) through the apical membrane of epithelial cells. Controls the reabsorption of sodium in kidney, colon, lung and sweat glands. Also plays a role in taste perception.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: SCNN1D
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR