Anti-SCNN1D Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Lieferant: Bioss
BS-10141R-HRP
BOSSBS-10141R-HRPEA 900 EUR
BOSSBS-10141R-HRP
Anti-SCNN1D Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
Sodium permeable non-voltage-sensitive ion channel inhibited by the diuretic amiloride. Mediates the electrodiffusion of the luminal sodium (and water, which follows osmotically) through the apical membrane of epithelial cells. Controls the reabsorption of sodium in kidney, colon, lung and sweat glands. Also plays a role in taste perception.

Recommended Dilutions: IHC-P: 1:100-500

Type: Primary
Antigen: SCNN1D
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR