Anti-RAD51 Rabbit Polyclonal Antibody
Lieferant: Bioss
BS-10145R
BOSSBS-10145REA 725 EUR
BOSSBS-10145R
Anti-RAD51 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: RAD51
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat