Anti-PDSS2 Rabbit Polyclonal Antibody (Cy3®)
Lieferant: Bioss
BS-10178R-CY3
BOSSBS-10178R-CY3EA 900 EUR
BOSSBS-10178R-CY3
Anti-PDSS2 Rabbit Polyclonal Antibody (Cy3®)
Antikörper
The protein encoded by this gene is an enzyme that synthesizes the prenyl side-chain of coenzyme Q, or ubiquinone, one of the key elements in the respiratory chain. The gene product catalyzes the formation of all trans-polyprenyl pyrophosphates from isopentyl diphosphate in the assembly of polyisoprenoid side chains, the first step in coenzyme Q biosynthesis. Defects in this gene are a cause of coenzyme Q10 deficiency.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: PDSS2
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Dog