Anti-CD5 Rabbit Polyclonal Antibody

Lieferant: Bioss
BS-10218R
BOSSBS-10218REA 752 EUR
BOSSBS-10218R
Anti-CD5 Rabbit Polyclonal Antibody
Antikörper
May act as a receptor in regulating T-cell proliferation.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: CD5
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR