Anti-CD93 Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Lieferant: Bioss
BS-10232R-A488
BOSSBS-10232R-A488EA 893 EUR
BOSSBS-10232R-A488
Anti-CD93 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antikörper
Receptor (or element of a larger receptor complex) for C1q, mannose-binding lectin (MBL2) and pulmonary surfactant protein A (SPA). May mediate the enhancement of phagocytosis in monocytes and macrophages upon interaction with soluble defense collagens. May play a role in intercellular adhesion.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CD93
Clonality: Polyclonal
Clone: 0
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR