Anti-CD93/C1qrp Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Lieferant: Bioss
BS-10232R-A680
BOSSBS-10232R-A680EA 890 EUR
BOSSBS-10232R-A680
Anti-CD93/C1qrp Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
Receptor (or element of a larger receptor complex) for C1q, mannose-binding lectin (MBL2) and pulmonary surfactant protein A (SPA). May mediate the enhancement of phagocytosis in monocytes and macrophages upon interaction with soluble defense collagens. May play a role in intercellular adhesion.

Type: Primary
Antigen: CD93/C1qrp
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 56-160/652
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR