Anti-L-Citrulline Rabbit Polyclonal Antibody (Cy7®)

Lieferant: Bioss
BS-10255R-CY7
BOSSBS-10255R-CY7EA 900 EUR
BOSSBS-10255R-CY7
Anti-L-Citrulline Rabbit Polyclonal Antibody (Cy7®)
Antikörper
The amino acid Citrulline is required to detoxify the liver from ammonia, which is a waste product of the body from oxidation. Citrulline promotes energy and assists with the immune system. This unusual amino acid is formed in the urea cycle by the addition of carbon dioxide and ammonia to ornithine. It is then combined with aspartic acid to form arginosuccinic acid, which later is metabolized into the amino acid arginine.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: L-Citrulline
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Others
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR