Anti-L-Citrulline Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Lieferant: Bioss
BS-10255R-HRP
BOSSBS-10255R-HRPEA 900 EUR
BOSSBS-10255R-HRP
Anti-L-Citrulline Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
The amino acid Citrulline is required to detoxify the liver from ammonia, which is a waste product of the body from oxidation. Citrulline promotes energy and assists with the immune system. This unusual amino acid is formed in the urea cycle by the addition of carbon dioxide and ammonia to ornithine. It is then combined with aspartic acid to form arginosuccinic acid, which later is metabolized into the amino acid arginine.
Recommended Dilutions: IHC-P: 1:100-500
Type: Primary
Antigen: L-Citrulline
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Others