Anti-LYZ Rabbit Polyclonal Antibody
Lieferant: Bioss
BS-10293R
BOSSBS-10293REA 725 EUR
BOSSBS-10293R
Anti-LYZ Rabbit Polyclonal Antibody
Antikörper
Lysozyme catalyzes the hydrolysis of certain mucopolysaccharides of bacterial cell walls. Specifically, it catalyzes the hydrolysis of the bacterial cell wall beta glycosidic linkages between N acetylmuramic acid and N acetylglucosamine. It is found in spleen, lung, kidney, white blood cells, plasma, saliva, milk, and tears.
IHC-P(1:100-500), IF(IHC-P)(1:50-200)
Type: Primary
Antigen: LYZ
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human