Anti-TN-C Rabbit Polyclonal Antibody (Cy3®)

Lieferant: Bioss
BS-1039R-CY3
BOSSBS-1039R-CY3EA 909 EUR
BOSSBS-1039R-CY3
Anti-TN-C Rabbit Polyclonal Antibody (Cy3®)
Antikörper
Extracellular matrix protein implicated in guidance of migrating neurons as well as axons during development, synaptic plasticity as well as neuronal regeneration. Promotes neurite outgrowth from cortical neurons grown on a monolayer of astrocytes. Ligand for integrins alpha-8/beta-1, alpha-9/beta-1, alpha-V/beta-3 and alpha-V/beta-6.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: Tenascin C
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR