Anti-VAMP1 / VAMP2 / VAMP3 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Lieferant: Bioss
BS-10427R-A350
BOSSBS-10427R-A350EA 893 EUR
BOSSBS-10427R-A350
Anti-VAMP1 / VAMP2 / VAMP3 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antikörper
Synaptobrevins/VAMPs, syntaxins, and the 25-kD synaptosomal-associated protein are the main components of a protein complex involved in the docking and/or fusion of synaptic vesicles with the presynaptic membrane. This gene is a member of the vesicle-associated membrane protein (VAMP)/synaptobrevin family. Because of its high homology to other known VAMPs, its broad tissue distribution, and its subcellular localization, the protein encoded by this gene was shown to be the human equivalent of the rodent cellubrevin. In platelets the protein resides on a compartment that is not mobilized toMore Product Information