Anti-VAMP-1+2+3 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Lieferant: Bioss
BS-10427R-A750
BOSSBS-10427R-A750EA 890 EUR
BOSSBS-10427R-A750
Anti-VAMP-1+2+3 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
Synaptobrevins/VAMPs, syntaxins, and the 25 kD synaptosomal-associated protein are the main components of a protein complex involved in the docking and/or fusion of synaptic vesicles with the presynaptic membrane. This gene is a member of the vesicle-associated membrane protein (VAMP)/synaptobrevin family. Because of its high homology to other known VAMPs, its broad tissue distribution, and its subcellular localisation, the protein encoded by this gene was shown to be the human equivalent of the rodent cellubrevin. In platelets the protein resides on a compartment that is not mobilized to the plasma membrane on calcium or thrombin stimulation.
Type: Primary
Antigen: VAMP-1+2+3
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 21-118/118
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen