Anti-PTGFR Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Lieferant: Bioss
BS-10433R-A647
BOSSBS-10433R-A647EA 893 EUR
BOSSBS-10433R-A647
Anti-PTGFR Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antikörper
Receptor for prostaglandin F2-alpha (PGF2-alpha). The activity of this receptor is mediated by G proteins which activate a phosphatidylinositol-calcium second messenger system. Initiates luteolysis in the corpus luteum (By similarity). Isoforms 2 to 7 do not bind PGF2-alpha but are proposed to modulate signaling by participating in variant receptor complexes; heterodimers between isoform 1 and isoform 5 are proposed to be a receptor for prostamides including the synthetic analog bimatoprost.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: PTGFR
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR