Anti-PTGFR Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Lieferant: Bioss
BS-10433R-A680
BOSSBS-10433R-A680EA 890 EUR
BOSSBS-10433R-A680
Anti-PTGFR Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
Receptor for prostaglandin F2-alpha (PGF2-alpha). The activity of this receptor is mediated by G proteins which activate a phosphatidylinositol-calcium second messenger system. Initiates luteolysis in the corpus luteum (By similarity). Isoforms 2 to 7 do not bind PGF2-alpha but are proposed to modulate Signalling by participating in variant receptor complexes; heterodimers between isoform 1 and isoform 5 are proposed to be a receptor for prostamides including the synthetic analog bimatoprost.
Type: Primary
Antigen: PTGFR
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 121-220/359
Host: Rabbit
Isotype: IgG
Reactivity: Human