Anti-PTGFR Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Lieferant: Bioss
BS-10433R-A750
BOSSBS-10433R-A750EA 890 EUR
BOSSBS-10433R-A750
Anti-PTGFR Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
Receptor for prostaglandin F2-alpha (PGF2-alpha). The activity of this receptor is mediated by G proteins which activate a phosphatidylinositol-calcium second messenger system. Initiates luteolysis in the corpus luteum (By similarity). Isoforms 2 to 7 do not bind PGF2-alpha but are proposed to modulate Signalling by participating in variant receptor complexes; heterodimers between isoform 1 and isoform 5 are proposed to be a receptor for prostamides including the synthetic analog bimatoprost.

Type: Primary
Antigen: PTGFR
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 121-220/359
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR