Anti-PTGFR Rabbit Polyclonal Antibody (Cy7®)
Lieferant: Bioss
BS-10433R-CY7
BOSSBS-10433R-CY7EA 900 EUR
BOSSBS-10433R-CY7
Anti-PTGFR Rabbit Polyclonal Antibody (Cy7®)
Antikörper
Receptor for prostaglandin F2-alpha (PGF2-alpha). The activity of this receptor is mediated by G proteins which activate a phosphatidylinositol-calcium second messenger system. Initiates luteolysis in the corpus luteum (By similarity). Isoforms 2 to 7 do not bind PGF2-alpha but are proposed to modulate signaling by participating in variant receptor complexes; heterodimers between isoform 1 and isoform 5 are proposed to be a receptor for prostamides including the synthetic analog bimatoprost.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: PTGFR
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat