Anti-NDUFS2 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Lieferant: Bioss
BS-10455R-A647
BOSSBS-10455R-A647EA 893 EUR
BOSSBS-10455R-A647
Anti-NDUFS2 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antikörper
Core subunit of the mitochondrial membrane respiratory chain NADH dehydrogenase (Complex I) that is believed to belong to the minimal assembly required for catalysis. Complex I functions in the transfer of electrons from NADH to the respiratory chain. The immediate electron acceptor for the enzyme is believed to be ubiquinone (By similarity).
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: NDUFS2
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat