Anti-RHOG Rabbit Polyclonal Antibody (Cy5®)

Lieferant: Bioss
BS-10466R-CY5
BOSSBS-10466R-CY5EA 900 EUR
BOSSBS-10466R-CY5
Anti-RHOG Rabbit Polyclonal Antibody (Cy5®)
Antikörper
This gene encodes a member of the Rho family of small GTPases, which cycle between inactive GDP-bound and active GTP-bound states and function as molecular switches in signal transduction cascades. Rho proteins promote reorganization of the actin cytoskeleton and regulate cell shape, attachment, and motility. The encoded protein facilitates translocation of a functional guanine nucleotide exchange factor (GEF) complex from the cytoplasm to the plasma membrane where ras-related C3 botulinum toxin substrate 1 is activated to promote lamellipodium formation and cell migration. Two related pseudogene have been identified on chromosomes 20 and X. [provided by RefSeq, Aug 2011].

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: RHOG
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR