Anti-VAP1 Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Lieferant: Bioss
BS-10529R-A680
BOSSBS-10529R-A680EA 890 EUR
BOSSBS-10529R-A680
Anti-VAP1 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
This gene encodes a member of the semicarbazide-sensitive amine oxidase family. Copper amine oxidases catalyse the oxidative conversion of amines to aldehydes in the presence of copper and quinone cofactor. The encoded protein is localised to the cell surface, has adhesive properties as well as monoamine oxidase activity, and may be involved in leukocyte trafficking. Alterations in levels of the encoded protein may be associated with many diseases, including diabetes mellitus. A pseudogene of this gene has been described and is located approximately 9-kb downstream on the same chromosome. Alternative splicing results in multiple transcript variants.

Type: Primary
Antigen: VAP1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 201-300/763
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR