Anti-AOC3 Rabbit Polyclonal Antibody (Cy7®)

Lieferant: Bioss
BS-10529R-CY7
BOSSBS-10529R-CY7EA 900 EUR
BOSSBS-10529R-CY7
Anti-AOC3 Rabbit Polyclonal Antibody (Cy7®)
Antikörper
This gene encodes a member of the semicarbazide-sensitive amine oxidase family. Copper amine oxidases catalyze the oxidative conversion of amines to aldehydes in the presence of copper and quinone cofactor. The encoded protein is localized to the cell surface, has adhesive properties as well as monoamine oxidase activity, and may be involved in leukocyte trafficking. Alterations in levels of the encoded protein may be associated with many diseases, including diabetes mellitus. A pseudogene of this gene has been described and is located approximately 9-kb downstream on the same chromosome. Alternative splicing results in multiple transcript variants. [provided by RefSeq, Apr 2013]

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: AOC3
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR