Anti-ITGA3 + ITGB1 Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Lieferant: Bioss
BS-1057R-A555
BOSSBS-1057R-A555EA 901 EUR
BOSSBS-1057R-A555
Anti-ITGA3 + ITGB1 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antikörper
alpha3beta1 integrin is a laminin receptor with apparently diverse functions. In epithelial cells it acts as a receptor for the basement membrane, whereas in neuronal and possibly tumor cells it mediates migration. Interactions of alpha3beta1 integrin with tetraspanin proteins may provide clues to how it transduces signals that affect cell behavior.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: Integrin Alpha 3 + Beta 1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR