Anti-ITGA3 + ITGB1 Rabbit Polyclonal Antibody (Cy3®)

Lieferant: Bioss
BS-1057R-CY3
BOSSBS-1057R-CY3EA 909 EUR
BOSSBS-1057R-CY3
Anti-ITGA3 + ITGB1 Rabbit Polyclonal Antibody (Cy3®)
Antikörper
alpha3beta1 integrin is a laminin receptor with apparently diverse functions. In epithelial cells it acts as a receptor for the basement membrane, whereas in neuronal and possibly tumor cells it mediates migration. Interactions of alpha3beta1 integrin with tetraspanin proteins may provide clues to how it transduces signals that affect cell behavior.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: Integrin Alpha 3 + Beta 1
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR