Anti-ITGA3 + ITGB1 Rabbit Polyclonal Antibody (Cy5®)
Lieferant: Bioss
BS-1057R-CY5
BOSSBS-1057R-CY5EA 909 EUR
BOSSBS-1057R-CY5
Anti-ITGA3 + ITGB1 Rabbit Polyclonal Antibody (Cy5®)
Antikörper
alpha3beta1 integrin is a laminin receptor with apparently diverse functions. In epithelial cells it acts as a receptor for the basement membrane, whereas in neuronal and possibly tumor cells it mediates migration. Interactions of alpha3beta1 integrin with tetraspanin proteins may provide clues to how it transduces signals that affect cell behavior.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: Integrin Alpha 3 + Beta 1
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat