Anti-ADRA2 Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Lieferant: Bioss
BS-1062R-A750
BOSSBS-1062R-A750EA 890 EUR
BOSSBS-1062R-A750
Anti-ADRA2 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
Alpha-2 adrenergic receptors mediate the catecholamine-induced inhibition of adenylate cyclase through the action of G proteins. The rank order of potency for agonists of this receptor is oxymetazoline > clonidine > epinephrine > norepinephrine > phenylephrine > dopamine > p-synephrine > p-tyramine > serotonin = p-octopamine. For antagonists, the rank order is yohimbine > phentolamine = mianserine > chlorpromazine = spiperone = prazosin > propanolol > alprenolol = pindolol.

Type: Primary
Antigen: ADRA2
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 385-450/450
Host: Rabbit
Isotype: IgG
Reactivity: Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR