Anti-ADRA2A Rabbit Polyclonal Antibody (Cy3®)

Lieferant: Bioss
BS-1062R-CY3
BOSSBS-1062R-CY3EA 893 EUR
BOSSBS-1062R-CY3
Anti-ADRA2A Rabbit Polyclonal Antibody (Cy3®)
Antikörper
Alpha-2 adrenergic receptors mediate the catecholamine-induced inhibition of adenylate cyclase through the action of G proteins. The rank order of potency for agonists of this receptor is oxymetazoline >clonidine >epinephrine >norepinephrine >phenylephrine >dopamine >p-synephrine >p-tyramine >serotonin = p-octopamine. For antagonists, the rank order is yohimbine >phentolamine = mianserine >chlorpromazine = spiperone = prazosin >propanolol >alprenolol = pindolol.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ADRA2A
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR